The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080670.3 | NP_542401.1 | 1 - 2 | 297 | 84 |
GenBank mRNA | AK307282.1 | - | 1 - 2 | 297 | 84 |