Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POMT1
Entrez Gene ID
10585
Gene Name
protein O-mannosyltransferase 1
Gene Aliases
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
Chromosome Location
Chr.9: 131502895 - 131523806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077365.1 NP_001070833.1 17 - 18 1905 87
NM_001077366.1 NP_001070834.1 16 - 17 1753 87
NM_001136113.1 NP_001129585.1 17 - 18 2145 87
NM_001136114.1 NP_001129586.1 15 - 16 1646 87
NM_007171.3 NP_009102.3 17 - 18 1971 87
XM_005272156.1 XP_005272213.1 17 - 18 2097 87
XM_005272158.1 XP_005272215.1 16 - 17 1826 87
XM_005272159.1 XP_005272216.1 15 - 16 1716 87
XM_005272162.2 XP_005272219.1 16 - 17 2147 87
XM_006716932.1 XP_006716995.1 16 - 17 1749 87
XM_011518140.1 XP_011516442.1 16 - 17 1824 87
XM_011518141.1 XP_011516443.1 16 - 17 1758 87
XM_011518142.1 XP_011516444.1 15 - 16 1626 87
XM_011518143.1 XP_011516445.1 15 - 16 1656 87
XM_011518145.1 XP_011516447.1 14 - 15 1492 87
XM_017014203.1 XP_016869692.1 17 - 18 2419 87
XM_017014204.1 XP_016869693.1 16 - 17 1693 87
XM_017014205.1 XP_016869694.1 17 - 18 1898 87
GenBank mRNA AB209430.1 - 11 - 12 3601 87
AF095136.1 - 17 - 18 1948 87
AK000475.1 - 6 - 7 560 87
AK074874.1 - 17 - 18 1863 87
AK074888.1 - 16 - 17 1737 87
AK094558.1 - 7 - 8 2253 87
AK293529.1 - 15 - 16 1558 87
AK295561.1 - 15 - 16 1650 87
AK299646.1 - 14 - 15 1457 87
AK300393.1 - 16 - 17 1773 87
AK315218.1 - 16 - 17 2108 87
BC022877.2 - 17 - 18 1799 87
BC065268.1 - 17 - 18 2090 87

View Full Product Details