The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277126.1 | NP_001264055.1 | 3 - 4 | 2303 | 118 |
XM_011527479.1 | XP_011525781.1 | 3 - 4 | 2303 | 118 | |
XM_011527480.1 | XP_011525782.1 | 3 - 4 | 2303 | 118 | |
XM_011527482.1 | XP_011525784.1 | 3 - 4 | 2303 | 118 | |
XM_017027460.1 | XP_016882949.1 | 3 - 4 | 2303 | 118 | |
XM_017027461.1 | XP_016882950.1 | 3 - 4 | 2303 | 118 | |
XM_017027463.1 | XP_016882952.1 | 4 - 5 | 2076 | 118 | |
XM_017027464.1 | XP_016882953.1 | 3 - 4 | 1874 | 118 | |
XM_017027465.1 | XP_016882954.1 | 4 - 5 | 2084 | 118 | |
XM_017027466.1 | XP_016882955.1 | 4 - 5 | 2081 | 118 | |
XM_017027467.1 | XP_016882956.1 | 3 - 4 | 1876 | 118 | |
GenBank mRNA | AY116204.1 | - | 3 - 4 | 2294 | 118 |
AY116205.1 | - | 3 - 4 | 2294 | 118 | |
AY116206.1 | - | 3 - 4 | 2294 | 118 | |
AY116207.1 | - | 3 - 4 | 2294 | 118 |