The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005191.2 | NP_001005191.1 | 1 - 1 | 944 | 132 |
GenBank mRNA | BC137147.1 | - | 1 - 1 | 944 | 132 |
BC137162.1 | - | 1 - 1 | 944 | 132 |