Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
Chromosome Location
Chr.11: 74696428 - 74731385 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278473.2 NP_001265402.1 3 - 4 784 58
NM_001304390.1 NP_001291319.1 2 - 3 676 58
NM_001304391.1 NP_001291320.1 2 - 3 676 58
NM_001304415.1 NP_001291344.1 2 - 3 676 58
NM_015424.5 NP_056239.3 3 - 4 784 58
GenBank mRNA AK304519.1 - 3 - 4 552 58
AY163868.1 - 3 - 4 290 58
AY279090.1 - 3 - 4 581 58
AY279091.1 - 3 - 4 581 58
AY279093.1 - 2 - 3 676 58
AY358522.1 - 3 - 4 573 58
BC142623.1 - 3 - 4 784 58
JN579714.1 - 3 - 4 287 58

View Full Product Details