Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL2A1
Entrez Gene ID
1280
Gene Name
collagen type II alpha 1 chain
Gene Aliases
ANFH, AOM, COL11A3, SEDC, STL1
Chromosome Location
Chr.12: 47972965 - 48006212 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001844.4 NP_001835.3 15 - 16 1152 76
NM_033150.2 NP_149162.2 14 - 15 945 76
XM_017018828.1 XP_016874317.1 17 - 18 1237 76
XM_017018829.1 XP_016874318.1 17 - 18 1234 76
XM_017018830.1 XP_016874319.1 16 - 17 1027 76
XM_017018831.1 XP_016874320.1 15 - 16 2537 76
GenBank mRNA BC116449.1 - 15 - 16 973 76
X16468.1 - 14 - 15 764 76
X16711.1 - 14 - 15 764 76

View Full Product Details