The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001844.4 | NP_001835.3 | 5 - 6 | 559 | 88 |
NM_033150.2 | NP_149162.2 | 4 - 5 | 352 | 88 | |
XM_017018828.1 | XP_016874317.1 | 7 - 8 | 644 | 88 | |
XM_017018829.1 | XP_016874318.1 | 7 - 8 | 641 | 88 | |
XM_017018830.1 | XP_016874319.1 | 6 - 7 | 434 | 88 | |
XM_017018831.1 | XP_016874320.1 | 5 - 6 | 1944 | 88 | |
GenBank mRNA | BC116449.1 | - | 5 - 6 | 380 | 88 |
X16468.1 | - | 4 - 5 | 171 | 88 | |
X16711.1 | - | 4 - 5 | 171 | 88 |