The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306190.1 | NP_001293119.1 | 1 - 2 | 344 | 156 |
NM_001698.2 | NP_001689.1 | 1 - 2 | 299 | 156 | |
XM_005252072.2 | XP_005252129.1 | 1 - 2 | 299 | 156 | |
XM_011518804.2 | XP_011517106.1 | 1 - 2 | 320 | 156 | |
XM_017014849.1 | XP_016870338.1 | 1 - 2 | 299 | 156 | |
GenBank mRNA | AK302453.1 | - | 1 - 2 | 270 | 156 |
BC020722.1 | - | 1 - 2 | 288 | 156 | |
BI667104.1 | - | 1 - 2 | 352 | 156 | |
DA724873.1 | - | 1 - 2 | 299 | 156 | |
X79888.1 | - | 1 - 2 | 268 | 156 |