The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001698.2 | NP_001689.1 | 3 - 4 | 454 | 119 |
XM_005252066.3 | XP_005252123.1 | 4 - 5 | 497 | 119 | |
XM_005252067.4 | XP_005252124.1 | 4 - 5 | 499 | 119 | |
XM_005252069.4 | XP_005252126.1 | 4 - 5 | 500 | 119 | |
XM_005252072.2 | XP_005252129.1 | 3 - 4 | 454 | 119 | |
XM_011518800.2 | XP_011517102.1 | 4 - 5 | 501 | 119 | |
XM_011518802.2 | XP_011517104.1 | 4 - 5 | 523 | 119 | |
XM_017014849.1 | XP_016870338.1 | 3 - 4 | 454 | 119 | |
XM_017014850.1 | XP_016870339.1 | 4 - 5 | 594 | 119 | |
XM_017014851.1 | XP_016870340.1 | 4 - 5 | 523 | 119 | |
GenBank mRNA | AK302453.1 | - | 3 - 4 | 425 | 119 |
BG706374.1 | - | 3 - 4 | 450 | 119 | |
X79888.1 | - | 3 - 4 | 423 | 119 |