The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306190.1 | NP_001293119.1 | 7 - 8 | 885 | 77 |
NM_001698.2 | NP_001689.1 | 8 - 9 | 927 | 77 | |
XM_005252066.3 | XP_005252123.1 | 9 - 10 | 970 | 77 | |
XM_006717150.3 | XP_006717213.1 | 8 - 9 | 886 | 77 | |
XM_011518802.2 | XP_011517104.1 | 9 - 10 | 996 | 77 | |
XM_017014850.1 | XP_016870339.1 | 9 - 10 | 1067 | 77 | |
GenBank mRNA | BC020722.1 | - | 7 - 8 | 829 | 77 |
X79888.1 | - | 8 - 9 | 896 | 77 |