The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005014.2 | NP_005005.1 | 2 - 3 | 1302 | 115 |
GenBank mRNA | AB000114.1 | - | 2 - 3 | 1033 | 115 |
AK225922.1 | - | 2 - 3 | 1147 | 115 | |
AK313055.1 | - | 2 - 3 | 1141 | 115 | |
AY358872.1 | - | 2 - 3 | 1200 | 115 | |
BC046356.1 | - | 2 - 3 | 1204 | 115 |