Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSC1
Entrez Gene ID
7248
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
Chromosome Location
Chr.9: 132891348 - 132945269 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000368.4 NP_000359.1 11 - 12 1376 73
NM_001162427.1 NP_001155899.1 10 - 11 1223 73
XM_005272211.1 XP_005272268.1 11 - 12 1421 73
XM_006717271.1 XP_006717334.1 11 - 12 1356 73
XM_011518979.2 XP_011517281.1 11 - 12 1370 73
XM_017015096.1 XP_016870585.1 12 - 13 1835 73
XM_017015097.1 XP_016870586.1 12 - 13 1806 73
XM_017015099.1 XP_016870588.1 10 - 11 1514 73
XM_017015100.1 XP_016870589.1 10 - 11 1485 73
GenBank mRNA AB190910.1 - 11 - 12 1338 73
AF013168.1 - 11 - 12 1363 73
AK303030.1 - 10 - 11 1206 73
KJ534953.1 - 9 - 10 1142 73

View Full Product Details