The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135671.2 | NP_001129143.1 | 2 - 3 | 564 | 85 |
NM_001199729.2 | NP_001186658.1 | 3 - 4 | 639 | 85 | |
NM_012331.4 | NP_036463.1 | 2 - 3 | 462 | 85 | |
XM_011543822.1 | XP_011542124.1 | 2 - 3 | 507 | 85 | |
XM_011543823.2 | XP_011542125.1 | 2 - 3 | 507 | 85 | |
XM_017013448.1 | XP_016868937.1 | 2 - 3 | 507 | 85 | |
XM_017013449.1 | XP_016868938.1 | 2 - 3 | 1268 | 85 | |
XM_017013450.1 | XP_016868939.1 | 2 - 3 | 205 | 85 | |
XM_017013451.1 | XP_016868940.1 | 2 - 3 | 1305 | 85 | |
XM_017013452.1 | XP_016868941.1 | 3 - 4 | 1369 | 85 | |
XM_017013453.1 | XP_016868942.1 | 3 - 4 | 1369 | 85 | |
XM_017013454.1 | XP_016868943.1 | 2 - 3 | 2679 | 85 | |
GenBank mRNA | AF086925.1 | - | 2 - 3 | 281 | 85 |
AF183420.1 | - | 2 - 3 | 271 | 85 | |
AJ242973.1 | - | 2 - 3 | 426 | 85 | |
AK293488.1 | - | 3 - 4 | 639 | 85 | |
AK299948.1 | - | 2 - 3 | 389 | 85 | |
AY690665.1 | - | 2 - 3 | 231 | 85 | |
AY958429.1 | - | 2 - 3 | 462 | 85 | |
AY958430.1 | - | 3 - 4 | 635 | 85 | |
AY958431.1 | - | 2 - 3 | 560 | 85 | |
BC054033.1 | - | 2 - 3 | 411 | 85 |