The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017845.3 | NP_060315.1 | 2 - 3 | 230 | 157 |
XM_017008330.1 | XP_016863819.1 | 2 - 3 | 273 | 157 | |
GenBank mRNA | AF182421.1 | - | 2 - 3 | 260 | 157 |
AK000509.1 | - | 2 - 3 | 250 | 157 | |
AY542163.1 | - | 2 - 3 | 222 | 157 | |
BC008371.1 | - | 2 - 3 | 290 | 157 | |
BC015145.1 | - | 2 - 3 | 234 | 157 | |
BC019826.1 | - | 2 - 3 | 256 | 157 |