Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PACSIN2
Entrez Gene ID
11252
Gene Name
protein kinase C and casein kinase substrate in neurons 2
Gene Aliases
SDPII
Chromosome Location
Chr.22: 42869766 - 43015690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184970.1 NP_001171899.1 6 - 7 1021 80
NM_001184971.1 NP_001171900.1 6 - 7 946 80
NM_007229.3 NP_009160.2 6 - 7 1008 80
XM_005261319.3 XP_005261376.1 6 - 7 1533 80
XM_011529846.1 XP_011528148.1 6 - 7 1533 80
XM_011529847.2 XP_011528149.1 6 - 7 1118 80
XM_011529848.2 XP_011528150.1 6 - 7 970 80
XM_011529849.2 XP_011528151.1 6 - 7 1154 80
XM_011529850.2 XP_011528152.1 6 - 7 1345 80
XM_011529851.2 XP_011528153.1 6 - 7 1035 80
XM_011529852.1 XP_011528154.1 5 - 6 1396 80
XM_011529853.1 XP_011528155.1 6 - 7 1533 80
XM_017028562.1 XP_016884051.1 6 - 7 999 80
XM_017028563.1 XP_016884052.1 6 - 7 1051 80
XM_017028564.1 XP_016884053.1 6 - 7 1109 80
XM_017028565.1 XP_016884054.1 6 - 7 1138 80
XM_017028566.1 XP_016884055.1 6 - 7 1174 80
XM_017028567.1 XP_016884056.1 6 - 7 1365 80
XM_017028568.1 XP_016884057.1 5 - 6 1396 80
GenBank mRNA AL136845.1 - 6 - 7 946 80
BC008037.2 - 6 - 7 1008 80
CR456536.1 - 5 - 6 858 80
CR533523.1 - 5 - 6 786 80

View Full Product Details