The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184970.1 | NP_001171899.1 | 9 - 10 | 1387 | 72 |
NM_007229.3 | NP_009160.2 | 9 - 10 | 1374 | 72 | |
XM_011529846.1 | XP_011528148.1 | 10 - 11 | 1905 | 72 | |
XM_011529847.2 | XP_011528149.1 | 10 - 11 | 1490 | 72 | |
XM_011529848.2 | XP_011528150.1 | 10 - 11 | 1342 | 72 | |
XM_011529849.2 | XP_011528151.1 | 10 - 11 | 1526 | 72 | |
XM_011529850.2 | XP_011528152.1 | 10 - 11 | 1717 | 72 | |
XM_011529851.2 | XP_011528153.1 | 10 - 11 | 1407 | 72 | |
XM_011529852.1 | XP_011528154.1 | 9 - 10 | 1768 | 72 | |
XM_017028562.1 | XP_016884051.1 | 10 - 11 | 1371 | 72 | |
XM_017028563.1 | XP_016884052.1 | 10 - 11 | 1423 | 72 | |
XM_017028564.1 | XP_016884053.1 | 9 - 10 | 1475 | 72 | |
GenBank mRNA | AF128536.1 | - | 9 - 10 | 1360 | 72 |
BC008037.2 | - | 9 - 10 | 1374 | 72 | |
CR456536.1 | - | 8 - 9 | 1224 | 72 |