The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011663.1 | NP_001011663.1 | 7 - 8 | 873 | 118 |
GenBank mRNA | AB047006.1 | - | 7 - 8 | 827 | 118 |
AK027885.1 | - | 7 - 8 | 849 | 118 | |
AK290679.1 | - | 7 - 8 | 827 | 118 | |
AK309105.1 | - | 8 - 9 | 1015 | 238 | |
BC010235.1 | - | 7 - 8 | 879 | 118 |