Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP63
Entrez Gene ID
80254
Gene Name
centrosomal protein 63
Gene Aliases
SCKL6
Chromosome Location
Chr.3: 134485733 - 134629607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042383.1 NP_001035842.1 9 - 10 1395 140
NM_001042384.1 NP_001035843.1 9 - 10 1395 140
NM_001042400.1 NP_001035859.1 10 - 11 1361 140
NM_025180.3 NP_079456.2 11 - 12 1637 140
XM_005247795.4 XP_005247852.1 10 - 11 1310 140
XM_005247797.3 XP_005247854.1 10 - 11 1565 140
XM_005247799.3 XP_005247856.1 10 - 11 1378 140
XM_006713760.3 XP_006713823.1 10 - 11 1242 140
XM_011513194.2 XP_011511496.1 10 - 11 1359 140
XM_011513197.2 XP_011511499.1 9 - 10 1193 140
XM_017007244.1 XP_016862733.1 10 - 11 1353 140
XM_017007245.1 XP_016862734.1 9 - 10 1212 140
XM_017007246.1 XP_016862735.1 9 - 10 1104 140
XM_017007247.1 XP_016862736.1 9 - 10 1221 140
XM_017007248.1 XP_016862737.1 9 - 10 1388 140
XM_017007249.1 XP_016862738.1 10 - 11 1477 140
XM_017007250.1 XP_016862739.1 10 - 11 1351 140
XM_017007251.1 XP_016862740.1 10 - 11 1349 140
XM_017007252.1 XP_016862741.1 10 - 11 1284 140
XM_017007253.1 XP_016862742.1 10 - 11 1345 140
XM_017007254.1 XP_016862743.1 10 - 11 1242 140
XM_017007255.1 XP_016862744.1 10 - 11 1565 140
XM_017007256.1 XP_016862745.1 11 - 12 1615 140
XM_017007257.1 XP_016862746.1 10 - 11 1347 140
XM_017007258.1 XP_016862747.1 10 - 11 1242 140
XM_017007259.1 XP_016862748.1 10 - 11 1359 140
XM_017007260.1 XP_016862749.1 9 - 10 1113 140
XM_017007261.1 XP_016862750.1 9 - 10 1172 140
XM_017007262.1 XP_016862751.1 9 - 10 1221 140
XM_017007263.1 XP_016862752.1 9 - 10 1427 140
XM_017007264.1 XP_016862753.1 10 - 11 1477 140
XM_017007265.1 XP_016862754.1 9 - 10 1205 140
XM_017007266.1 XP_016862755.1 9 - 10 1221 140
XM_017007267.1 XP_016862756.1 9 - 10 1388 140
XM_017007268.1 XP_016862757.1 9 - 10 1392 140
XM_017007269.1 XP_016862758.1 10 - 11 1495 140
GenBank mRNA AF113697.1 - 2 - 3 140
AK023448.1 - 10 - 11 1307 140
AK023738.1 - 9 - 10 1336 140
AK056465.1 - 11 - 12 1615 140
BC014050.2 - 9 - 10 1384 140
BC068997.1 - 10 - 11 1281 140

View Full Product Details