Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
Chromosome Location
Chr.14: 100376410 - 100530303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011537150.2 XP_011535452.1 1 - 2 336 96

View Full Product Details