Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
Chromosome Location
Chr.14: 100376410 - 100530303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161476.1 NP_001154948.1 6 - 7 1503 52
NM_024515.4 NP_078791.3 6 - 7 1634 52
XM_005268056.4 XP_005268113.1 6 - 7 1510 52
XM_005268057.3 XP_005268114.1 5 - 6 660 52
XM_005268058.4 XP_005268115.1 5 - 6 674 52
XM_011537150.2 XP_011535452.1 6 - 7 1757 52
XM_011537151.2 XP_011535453.2 5 - 6 1113 52
XM_017021639.1 XP_016877128.1 5 - 6 662 52
GenBank mRNA AK291960.1 - 6 - 7 1466 52
AY295082.1 - 6 - 7 1517 52
BC003641.2 - 5 - 6 740 52
BC007953.2 - 4 - 5 1287 52

View Full Product Details