The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152688.3 | NP_689901.2 | 4 - 5 | 761 | 81 |
XM_011535515.2 | XP_011533817.1 | 4 - 5 | 724 | 81 | |
XM_011535516.1 | XP_011533818.1 | 4 - 5 | 724 | 81 | |
XM_011535517.1 | XP_011533819.1 | 4 - 5 | 724 | 81 | |
XM_011535519.1 | XP_011533821.1 | 4 - 5 | 724 | 81 | |
XM_011535520.1 | XP_011533822.1 | 4 - 5 | 724 | 81 | |
XM_017010334.1 | XP_016865823.1 | 4 - 5 | 724 | 81 | |
XM_017010335.1 | XP_016865824.1 | 4 - 5 | 724 | 81 | |
XM_017010336.1 | XP_016865825.1 | 4 - 5 | 724 | 81 | |
XM_017010337.1 | XP_016865826.1 | 4 - 5 | 724 | 81 | |
XM_017010339.1 | XP_016865828.1 | 4 - 5 | 724 | 81 | |
XM_017010340.1 | XP_016865829.1 | 4 - 5 | 724 | 81 | |
XM_017010341.1 | XP_016865830.1 | 4 - 5 | 724 | 81 | |
XM_017010342.1 | XP_016865831.1 | 4 - 5 | 724 | 81 | |
XM_017010343.1 | XP_016865832.1 | 4 - 5 | 724 | 81 | |
XM_017010345.1 | XP_016865834.1 | 4 - 5 | 724 | 81 | |
XM_017010347.1 | XP_016865836.1 | 4 - 5 | 647 | 81 | |
GenBank mRNA | AK095983.1 | - | 4 - 5 | 709 | 81 |
AK292043.1 | - | 4 - 5 | 708 | 81 | |
BC034043.1 | - | 4 - 5 | 761 | 81 |