The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079673.1 | NP_001073141.1 | 15 - 16 | 2064 | 92 |
NM_001278438.1 | NP_001265367.1 | 15 - 16 | 2107 | 92 | |
NM_014923.4 | NP_055738.3 | 13 - 14 | 1688 | 92 | |
NR_103528.1 | 15 - 16 | 2107 | 92 | ||
XM_011534997.2 | XP_011533299.1 | 14 - 15 | 1778 | 92 | |
XM_017020440.1 | XP_016875929.1 | 15 - 16 | 1949 | 92 | |
XM_017020441.1 | XP_016875930.1 | 17 - 18 | 2570 | 92 | |
XM_017020442.1 | XP_016875931.1 | 16 - 17 | 2102 | 92 | |
XM_017020443.1 | XP_016875932.1 | 15 - 16 | 2422 | 92 | |
GenBank mRNA | AB023187.2 | - | 13 - 14 | 1618 | 92 |
AJ749706.1 | - | 15 - 16 | 2066 | 92 | |
AJ749707.1 | - | 13 - 14 | 1679 | 92 | |
AK302415.1 | - | 13 - 14 | 1688 | 92 | |
BC060816.1 | - | 6 - 7 | 726 | 92 | |
BC070072.1 | - | 6 - 7 | 726 | 92 | |
BC107793.1 | - | 12 - 13 | 1555 | 92 | |
BC132812.1 | - | 15 - 16 | 1831 | 92 | |
BC136617.1 | - | 15 - 16 | 1832 | 92 | |
BC144301.1 | - | 15 - 16 | 1832 | 92 | |
BX640739.1 | - | 15 - 16 | 1967 | 92 | |
BX648141.1 | - | 15 - 16 | 2054 | 92 | |
BX648706.1 | - | 17 - 18 | 1912 | 92 |