Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRLR
Entrez Gene ID
5618
Gene Name
prolactin receptor
Gene Aliases
HPRL, MFAB, hPRLrI
Chromosome Location
Chr.5: 35048295 - 35230589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000949.6 NP_000940.1 8 - 9 1211 58
NM_001204314.2 NP_001191243.1 5 - 6 586 58
NM_001204315.1 NP_001191244.1 7 - 8 889 58
NM_001204316.1 NP_001191245.1 7 - 8 889 58
NM_001204317.1 NP_001191246.1 7 - 8 889 58
NR_037910.1 6 - 7 756 58
XM_006714484.2 XP_006714547.1 8 - 9 1029 58
XM_011514068.2 XP_011512370.1 8 - 9 1044 58
XM_011514069.2 XP_011512371.1 8 - 9 983 58
XM_017009645.1 XP_016865134.1 7 - 8 977 58
XM_017009646.1 XP_016865135.1 6 - 7 827 58
GenBank mRNA AF166329.1 - 6 - 7 784 58
AF349939.1 - 4 - 5 481 58
AF416618.1 - 6 - 7 801 58
AF416619.1 - 6 - 7 801 58
AF493069.1 - 5 - 6 668 58
AF512796.1 - 5 - 6 475 58
AK313270.1 - 8 - 9 1099 58
BC059392.1 - 8 - 9 1097 58
GU133399.1 - 6 - 7 801 58
M31661.1 - 8 - 9 1068 58

View Full Product Details