The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001200053.1 | NP_001186982.1 | 7 - 8 | 1559 | 77 |
NM_001320122.1 | NP_001307051.1 | 10 - 11 | 1691 | 77 | |
NM_006928.4 | NP_008859.1 | 10 - 11 | 1852 | 77 | |
XM_006719569.1 | XP_006719632.1 | 9 - 10 | 2429 | 77 | |
GenBank mRNA | AK092881.1 | - | 9 - 10 | 2430 | 77 |
AK130180.1 | - | 10 - 11 | 1876 | 77 | |
AK300150.1 | - | 7 - 8 | 1539 | 77 | |
BC001414.2 | - | 9 - 10 | 1776 | 77 | |
BT007202.1 | - | 9 - 10 | 1766 | 77 | |
CA432578.1 | - | 2 - 3 | 77 | ||
M32295.1 | - | 9 - 10 | 1787 | 77 | |
S73003.1 | - | 9 - 10 | 1787 | 77 | |
U01874.1 | - | 9 - 10 | 80 |