Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GULP1
- Entrez Gene ID
-
51454
- Gene Name
- GULP, engulfment adaptor PTB domain containing 1
- Gene Aliases
-
CED-6, CED6, GULP
- Chromosome Location
-
Chr.2: 188291669 - 188595926 on Build GRCh38
View Full Product Details