Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCO4
Entrez Gene ID
255104
Gene Name
transmembrane and coiled-coil domains 4
Gene Aliases
-
Chromosome Location
Chr.1: 19682213 - 19800385 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181719.4 NP_859070.3 12 - 13 1423 68
XM_005245822.4 XP_005245879.1 12 - 13 1439 68
XM_005245824.4 XP_005245881.1 12 - 13 1503 68
XM_005245825.4 XP_005245882.1 11 - 12 1304 68
XM_005245827.3 XP_005245884.1 11 - 12 1319 68
XM_006710551.3 XP_006710614.1 12 - 13 1528 68
XM_011541179.2 XP_011539481.1 15 - 16 1898 68
XM_011541180.2 XP_011539482.1 15 - 16 2053 68
XM_011541181.1 XP_011539483.1 14 - 15 1727 68
XM_011541185.2 XP_011539487.1 12 - 13 1446 68
XM_011541186.2 XP_011539488.1 12 - 13 1543 68
XM_017000913.1 XP_016856402.1 11 - 12 1327 68
XM_017000914.1 XP_016856403.1 14 - 15 1947 68
XM_017000915.1 XP_016856404.1 14 - 15 1669 68
XM_017000916.1 XP_016856405.1 12 - 13 1581 68
XM_017000917.1 XP_016856406.1 13 - 14 1796 68
XM_017000918.1 XP_016856407.1 14 - 15 1993 68
XM_017000919.1 XP_016856408.1 13 - 14 1720 68
XM_017000920.1 XP_016856409.1 12 - 13 1938 68
XM_017000921.1 XP_016856410.1 13 - 14 1745 68
XM_017000922.1 XP_016856411.1 11 - 12 1404 68
GenBank mRNA AL049846.1 - 9 - 10 1189 68
AL049847.1 - 8 - 9 1067 68
AX775875.1 - 11 - 12 1452 68
AX775877.1 - 10 - 11 1332 68
BC053600.1 - 10 - 11 1282 68
BX640987.1 - 12 - 13 1424 68

View Full Product Details