The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136113.1 | NP_001129585.1 | 1 - 2 | 409 | 105 |
XM_005272156.1 | XP_005272213.1 | 1 - 2 | 295 | 105 | |
XM_017014203.1 | XP_016869692.1 | 1 - 2 | 308 | 105 | |
XM_017014205.1 | XP_016869694.1 | 1 - 2 | 91 | 105 | |
GenBank mRNA | BC065268.1 | - | 1 - 2 | 354 | 105 |