The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077365.1 | NP_001070833.1 | 8 - 9 | 894 | 61 |
NM_001077366.1 | NP_001070834.1 | 7 - 8 | 742 | 61 | |
NM_001136113.1 | NP_001129585.1 | 8 - 9 | 1134 | 61 | |
NM_001136114.1 | NP_001129586.1 | 6 - 7 | 635 | 61 | |
XM_011518141.1 | XP_011516443.1 | 7 - 8 | 747 | 61 | |
XM_011518143.1 | XP_011516445.1 | 6 - 7 | 640 | 61 | |
XM_017014204.1 | XP_016869693.1 | 7 - 8 | 677 | 61 | |
GenBank mRNA | AK074874.1 | - | 8 - 9 | 852 | 61 |
AK074888.1 | - | 7 - 8 | 726 | 61 | |
AK293529.1 | - | 6 - 7 | 547 | 61 | |
AK295561.1 | - | 6 - 7 | 639 | 61 | |
AK299646.1 | - | 5 - 6 | 446 | 61 | |
AK300393.1 | - | 7 - 8 | 762 | 61 | |
BC022877.2 | - | 8 - 9 | 788 | 61 | |
BC065268.1 | - | 8 - 9 | 1079 | 61 |