Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIABLO
Entrez Gene ID
56616
Gene Name
diablo IAP-binding mitochondrial protein
Gene Aliases
DFNA64, SMAC
Chromosome Location
Chr.12: 122207662 - 122227534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278342.1 NP_001265271.1 2 - 3 740 110
GenBank mRNA AK057778.1 - 2 - 3 213 110
AY313210.1 - 2 - 3 186 110
BF724951.1 - 2 - 3 207 110
KU178726.1 - 2 - 3 186 110

View Full Product Details