Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4E2
Entrez Gene ID
9470
Gene Name
eukaryotic translation initiation factor 4E family member 2
Gene Aliases
4E-LP, 4EHP, EIF4EL3, IF4e
Chromosome Location
Chr.2: 232550587 - 232583645 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276336.1 NP_001263265.1 1 - 2 152 57
NM_001276337.1 NP_001263266.1 1 - 2 152 57
NM_001282958.1 NP_001269887.1 1 - 2 152 57
NM_004846.3 NP_004837.1 1 - 2 152 57
XM_005246975.3 XP_005247032.1 1 - 2 103 57
XM_017005377.1 XP_016860866.1 1 - 2 103 57
GenBank mRNA AB209519.1 - 1 - 2 56 57
AF038957.1 - 1 - 2 44 57
AF047695.1 - 1 - 2 43 57
AF068117.1 - 1 - 2 28 57
AK022518.1 - 1 - 2 50 57
AK303802.1 - 1 - 2 50 57
BC005392.1 - 1 - 2 50 57
BC005874.2 - 1 - 2 72 57
BC021690.1 - 1 - 2 54 57
BG913475.1 - 1 - 2 53 57
CB994873.1 - - 120 57
CD684451.1 - 1 - 2 75 57
DA992544.1 - 1 - 2 152 57

View Full Product Details