Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35A5
Entrez Gene ID
55032
Gene Name
solute carrier family 35 member A5
Gene Aliases
-
Chromosome Location
Chr.3: 112561709 - 112584439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017945.3 NP_060415.1 2 - 3 410 83
XM_011512948.1 XP_011511250.1 2 - 3 260 83
GenBank mRNA AK000737.1 - 2 - 3 313 83
AK098839.1 - 2 - 3 323 83
AK297411.1 - 2 - 3 310 83
AY358994.1 - 2 - 3 319 83
BC005207.1 - 2 - 3 372 83
BC010307.1 - 2 - 3 372 83
BC013046.1 - 2 - 3 314 83
DA854399.1 - 2 - 3 411 83

View Full Product Details