The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204318.1 | NP_001191247.1 | 6 - 7 | 793 | 65 |
GenBank mRNA | AF492470.1 | - | 5 - 6 | 705 | 65 |
AF493068.1 | - | 4 - 5 | 572 | 65 |