Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC8A1
Entrez Gene ID
6546
Gene Name
solute carrier family 8 member A1
Gene Aliases
NCX1
Chromosome Location
Chr.2: 40112146 - 40512452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112800.1 NP_001106271.1 8 - 9 2557 92
NM_001112801.1 NP_001106272.1 7 - 8 2533 92
NM_001112802.1 NP_001106273.1 7 - 8 2551 92
NM_001252624.1 NP_001239553.1 8 - 9 2548 92
NM_021097.2 NP_066920.1 9 - 10 2572 92
XM_005264514.3 XP_005264571.1 7 - 8 2646 92
XM_006712081.2 XP_006712144.1 10 - 11 2754 92
XM_006712082.3 XP_006712145.1 11 - 12 2856 92
XM_006712083.3 XP_006712146.1 10 - 11 2673 92
XM_006712084.3 XP_006712147.1 11 - 12 2954 92
XM_006712085.3 XP_006712148.1 10 - 11 3576 92
XM_011533050.1 XP_011531352.1 8 - 9 2548 92
XM_011533054.2 XP_011531356.1 10 - 11 2590 92
XM_011533055.2 XP_011531357.1 10 - 11 2717 92
XM_011533056.1 XP_011531358.1 10 - 11 2590 92
XM_011533057.2 XP_011531359.2 7 - 8 2633 92
XM_017004745.1 XP_016860234.1 10 - 11 2738 92
XM_017004746.1 XP_016860235.1 10 - 11 2591 92
XM_017004747.1 XP_016860236.1 8 - 9 2757 92
XM_017004748.1 XP_016860237.1 10 - 11 2659 92
XM_017004749.1 XP_016860238.1 11 - 12 2859 92
XM_017004750.1 XP_016860239.1 12 - 13 3093 92
XM_017004751.1 XP_016860240.1 12 - 13 3090 92
XM_017004752.1 XP_016860241.1 11 - 12 2894 92
XM_017004753.1 XP_016860242.1 10 - 11 2590 92
XM_017004754.1 XP_016860243.1 9 - 10 2694 92
XM_017004755.1 XP_016860244.1 8 - 9 2681 92
XM_017004756.1 XP_016860245.1 8 - 9 2761 92
XM_017004757.1 XP_016860246.1 8 - 9 2756 92
XM_017004758.1 XP_016860247.1 8 - 9 2735 92
XM_017004759.1 XP_016860248.1 8 - 9 2696 92
XM_017004760.1 XP_016860249.1 8 - 9 2670 92
XM_017004761.1 XP_016860250.1 8 - 9 2713 92
XM_017004762.1 XP_016860251.1 7 - 8 2667 92
XM_017004763.1 XP_016860252.1 7 - 8 2649 92
GenBank mRNA AB209075.1 - 8 - 9 862 92
AF108388.1 - 7 - 8 2509 92
AF108389.1 - 6 - 7 2440 92
AF128524.1 - 7 - 8 2531 92
AK291696.1 - 7 - 8 2551 92
BC098285.1 - 9 - 10 782 92
BC098308.1 - 8 - 9 766 92
BX648299.1 - 7 - 8 2631 92
M91368.1 - - 2628 92
M96368.1 - 9 - 10 2589 92

View Full Product Details