Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC8A1
- Entrez Gene ID
-
6546
- Gene Name
- solute carrier family 8 member A1
- Gene Aliases
-
NCX1
- Chromosome Location
-
Chr.2: 40112146 - 40512452 on Build GRCh38
View Full Product Details