The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000231.2 | NP_000222.1 | 1 - 2 | 143 | 76 |
GenBank mRNA | BC074777.2 | - | 1 - 2 | 31 | 76 |
BC074778.2 | - | 1 - 2 | 30 | 76 | |
BC109321.2 | - | 1 - 2 | 103 | 76 | |
DA897096.1 | - | 1 - 2 | 59 | 76 | |
U34976.1 | - | 1 - 2 | 112 | 76 |