The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039582.3 | NP_001034671.3 | 7 - 8 | 1051 | 64 |
NM_001135740.1 | NP_001129212.1 | 7 - 8 | 685 | 64 | |
XM_005274651.2 | XP_005274708.1 | 7 - 8 | 668 | 64 | |
XM_006724809.3 | XP_006724872.1 | 7 - 8 | 877 | 64 | |
XM_011531107.2 | XP_011529409.2 | 7 - 8 | 756 | 64 | |
XM_011531108.2 | XP_011529410.1 | 7 - 8 | 721 | 64 | |
XM_011531110.2 | XP_011529412.1 | 7 - 8 | 773 | 64 | |
XM_011531111.2 | XP_011529413.1 | 7 - 8 | 691 | 64 | |
XM_011531112.2 | XP_011529414.1 | 7 - 8 | 664 | 64 | |
XM_017029277.1 | XP_016884766.1 | 7 - 8 | 976 | 64 | |
XM_017029278.1 | XP_016884767.1 | 8 - 9 | 880 | 64 | |
XM_017029279.1 | XP_016884768.1 | 7 - 8 | 637 | 64 | |
GenBank mRNA | AK296165.1 | - | 6 - 7 | 1094 | 64 |
AK296203.1 | - | 7 - 8 | 1051 | 64 | |
AK303746.1 | - | 7 - 8 | 685 | 64 | |
BC064422.1 | - | 6 - 7 | 743 | 64 |