The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001138.1 | NP_001129.1 | 2 - 3 | 431 | 95 |
GenBank mRNA | BC110443.2 | - | 1 - 2 | 134 | 95 |
U88063.1 | - | 2 - 3 | 431 | 95 | |
U89485.1 | - | 1 - 2 | 131 | 95 |