Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAPGEF1
Entrez Gene ID
2889
Gene Name
Rap guanine nucleotide exchange factor 1
Gene Aliases
C3G, GRF2
Chromosome Location
Chr.9: 131576770 - 131740068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304275.1 NP_001291204.1 14 - 15 2301 82
NM_005312.3 NP_005303.2 14 - 15 2260 82
NM_198679.1 NP_941372.1 14 - 15 2349 82
XM_005272186.4 XP_005272243.1 17 - 18 2935 82
XM_005272191.3 XP_005272248.1 16 - 17 2778 82
XM_006717067.3 XP_006717130.1 17 - 18 2897 82
XM_006717072.3 XP_006717135.1 16 - 17 2793 82
XM_006717074.3 XP_006717137.1 14 - 15 2387 82
XM_011518569.2 XP_011516871.1 17 - 18 3022 82
XM_011518570.2 XP_011516872.1 17 - 18 2911 82
XM_011518571.2 XP_011516873.1 17 - 18 3582 82
XM_011518572.2 XP_011516874.1 17 - 18 2761 82
XM_011518573.2 XP_011516875.1 17 - 18 2894 82
XM_011518574.2 XP_011516876.1 16 - 17 2821 82
XM_011518575.2 XP_011516877.1 16 - 17 2818 82
XM_011518576.2 XP_011516878.1 15 - 16 2563 82
XM_011518577.2 XP_011516879.1 15 - 16 2560 82
XM_011518578.2 XP_011516880.1 15 - 16 2497 82
XM_011518579.2 XP_011516881.1 14 - 15 2404 82
XM_017014633.1 XP_016870122.1 17 - 18 2907 82
XM_017014634.1 XP_016870123.1 17 - 18 2758 82
XM_017014635.1 XP_016870124.1 17 - 18 2812 82
XM_017014636.1 XP_016870125.1 17 - 18 2783 82
XM_017014637.1 XP_016870126.1 16 - 17 2755 82
XM_017014638.1 XP_016870127.1 15 - 16 2662 82
XM_017014639.1 XP_016870128.1 16 - 17 2656 82
GenBank mRNA AK302763.1 - 6 - 7 839 82
BC041710.1 - 14 - 15 2228 82
CR749354.1 - 14 - 15 2202 82
D21239.1 - 14 - 15 2266 82

View Full Product Details