Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAPGEF1
Entrez Gene ID
2889
Gene Name
Rap guanine nucleotide exchange factor 1
Gene Aliases
C3G, GRF2
Chromosome Location
Chr.9: 131576770 - 131740068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304275.1 NP_001291204.1 21 - 22 3020 59
NM_005312.3 NP_005303.2 21 - 22 2979 59
NM_198679.1 NP_941372.1 21 - 22 3068 59
XM_005272186.4 XP_005272243.1 24 - 25 3654 59
XM_005272191.3 XP_005272248.1 23 - 24 3497 59
XM_006717067.3 XP_006717130.1 24 - 25 3616 59
XM_006717072.3 XP_006717135.1 23 - 24 3512 59
XM_006717074.3 XP_006717137.1 21 - 22 3106 59
XM_011518569.2 XP_011516871.1 24 - 25 3741 59
XM_011518570.2 XP_011516872.1 24 - 25 3630 59
XM_011518571.2 XP_011516873.1 24 - 25 4301 59
XM_011518572.2 XP_011516874.1 24 - 25 3480 59
XM_011518573.2 XP_011516875.1 24 - 25 3613 59
XM_011518574.2 XP_011516876.1 23 - 24 3540 59
XM_011518575.2 XP_011516877.1 23 - 24 3537 59
XM_011518576.2 XP_011516878.1 22 - 23 3282 59
XM_011518577.2 XP_011516879.1 22 - 23 3279 59
XM_011518578.2 XP_011516880.1 22 - 23 3216 59
XM_011518579.2 XP_011516881.1 21 - 22 3123 59
XM_017014633.1 XP_016870122.1 24 - 25 3626 59
XM_017014634.1 XP_016870123.1 24 - 25 3477 59
XM_017014635.1 XP_016870124.1 24 - 25 3531 59
XM_017014636.1 XP_016870125.1 24 - 25 3502 59
XM_017014637.1 XP_016870126.1 23 - 24 3474 59
XM_017014638.1 XP_016870127.1 22 - 23 3381 59
XM_017014639.1 XP_016870128.1 23 - 24 3375 59
GenBank mRNA AK302763.1 - 13 - 14 1558 59
BC041710.1 - 21 - 22 2947 59
CR749354.1 - 21 - 22 2921 59
D21239.1 - 21 - 22 2985 59

View Full Product Details