Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAPGEF1
Entrez Gene ID
2889
Gene Name
Rap guanine nucleotide exchange factor 1
Gene Aliases
C3G, GRF2
Chromosome Location
Chr.9: 131576770 - 131740068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304275.1 NP_001291204.1 6 - 7 859 63
NM_005312.3 NP_005303.2 6 - 7 818 63
NM_198679.1 NP_941372.1 6 - 7 907 63
XM_005272186.4 XP_005272243.1 6 - 7 983 63
XM_005272191.3 XP_005272248.1 5 - 6 826 63
XM_006717067.3 XP_006717130.1 6 - 7 945 63
XM_006717072.3 XP_006717135.1 5 - 6 841 63
XM_006717074.3 XP_006717137.1 6 - 7 945 63
XM_011518569.2 XP_011516871.1 6 - 7 1070 63
XM_011518570.2 XP_011516872.1 6 - 7 962 63
XM_011518571.2 XP_011516873.1 6 - 7 1630 63
XM_011518572.2 XP_011516874.1 6 - 7 809 63
XM_011518573.2 XP_011516875.1 6 - 7 942 63
XM_011518574.2 XP_011516876.1 6 - 7 962 63
XM_011518575.2 XP_011516877.1 6 - 7 962 63
XM_011518576.2 XP_011516878.1 6 - 7 962 63
XM_011518577.2 XP_011516879.1 6 - 7 962 63
XM_011518578.2 XP_011516880.1 6 - 7 962 63
XM_011518579.2 XP_011516881.1 6 - 7 962 63
XM_011518580.2 XP_011516882.1 6 - 7 962 63
XM_011518581.2 XP_011516883.1 6 - 7 962 63
XM_011518582.2 XP_011516884.1 6 - 7 962 63
XM_017014633.1 XP_016870122.1 6 - 7 955 63
XM_017014634.1 XP_016870123.1 6 - 7 806 63
XM_017014635.1 XP_016870124.1 6 - 7 860 63
XM_017014636.1 XP_016870125.1 6 - 7 831 63
XM_017014637.1 XP_016870126.1 6 - 7 962 63
XM_017014638.1 XP_016870127.1 6 - 7 962 63
XM_017014639.1 XP_016870128.1 6 - 7 962 63
XM_017014640.1 XP_016870129.1 6 - 7 962 63
XM_017014641.1 XP_016870130.1 6 - 7 962 63
GenBank mRNA BC041710.1 - 6 - 7 786 63
CR749354.1 - 6 - 7 760 63
D21239.1 - 6 - 7 824 63

View Full Product Details