The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004655.3 | NP_004646.3 | 6 - 7 | 2002 | 68 |
XM_011525319.2 | XP_011523621.1 | 6 - 7 | 2213 | 68 | |
XM_011525320.1 | XP_011523622.1 | 6 - 7 | 2202 | 68 | |
XM_011525321.2 | XP_011523623.1 | 6 - 7 | 2150 | 68 | |
XM_017025192.1 | XP_016880681.1 | 6 - 7 | 1869 | 68 | |
GenBank mRNA | AF078165.1 | - | 5 - 6 | 1802 | 68 |
AK025718.1 | - | 5 - 6 | 1024 | 68 | |
AK315303.1 | - | 6 - 7 | 1918 | 68 | |
BC101533.1 | - | 5 - 6 | 1782 | 68 | |
CR933657.1 | - | 6 - 7 | 2027 | 68 | |
DA839648.1 | - | 1 - 2 | 480 | 68 |