The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016255.2 | NP_057339.1 | 4 - 5 | 1221 | 108 |
GenBank mRNA | AF097027.1 | - | 4 - 5 | 1153 | 108 |
AK296393.1 | - | 5 - 6 | 535 | 108 | |
AK307681.1 | - | 4 - 5 | 713 | 108 | |
AK312814.1 | - | 4 - 5 | 1156 | 108 | |
BC047881.1 | - | 4 - 5 | 1145 | 108 |