The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304548.1 | NP_001291477.1 | 11 - 12 | 2035 | 85 |
NM_152632.3 | NP_689845.2 | 11 - 12 | 2035 | 85 | |
XM_017029452.1 | XP_016884941.1 | 11 - 12 | 2028 | 85 | |
XM_017029453.1 | XP_016884942.1 | 11 - 12 | 2028 | 85 | |
GenBank mRNA | AK126295.1 | - | 11 - 12 | 2001 | 85 |
BC027936.1 | - | 11 - 12 | 1988 | 85 | |
DV080631.1 | - | - | 345 | 85 |