The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306190.1 | NP_001293119.1 | 3 - 4 | 496 | 74 |
XM_006717150.3 | XP_006717213.1 | 4 - 5 | 497 | 74 | |
GenBank mRNA | BC020722.1 | - | 3 - 4 | 440 | 74 |