The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193321.1 | NP_001180250.1 | 13 - 14 | 1965 | 75 |
NM_001193322.1 | NP_001180251.1 | 14 - 15 | 2084 | 75 | |
NM_014002.3 | NP_054721.1 | 14 - 15 | 2084 | 75 | |
XM_005273356.2 | XP_005273413.1 | 14 - 15 | 2080 | 75 | |
XM_017002868.1 | XP_016858357.1 | 13 - 14 | 2080 | 75 | |
GenBank mRNA | AB016590.1 | - | 12 - 13 | 1508 | 75 |
AF241789.1 | - | 12 - 13 | 1506 | 75 | |
AK093798.1 | - | 8 - 9 | 945 | 75 | |
BC105923.1 | - | 14 - 15 | 1832 | 75 | |
BC105924.1 | - | 14 - 15 | 1832 | 75 | |
BC107591.1 | - | 14 - 15 | 1832 | 75 | |
BC107812.1 | - | 13 - 14 | 1713 | 75 | |
D63485.1 | - | 14 - 15 | 1832 | 75 |