The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193321.1 | NP_001180250.1 | 5 - 6 | 999 | 57 |
NM_001193322.1 | NP_001180251.1 | 6 - 7 | 1118 | 57 | |
NM_014002.3 | NP_054721.1 | 6 - 7 | 1118 | 57 | |
XM_005273356.2 | XP_005273413.1 | 6 - 7 | 1114 | 57 | |
XM_017002868.1 | XP_016858357.1 | 5 - 6 | 1114 | 57 | |
GenBank mRNA | AB016590.1 | - | 4 - 5 | 542 | 57 |
AF241789.1 | - | 4 - 5 | 540 | 57 | |
AK310737.1 | - | 6 - 7 | 885 | 57 | |
BC105923.1 | - | 6 - 7 | 866 | 57 | |
BC107591.1 | - | 6 - 7 | 866 | 57 | |
BC107812.1 | - | 5 - 6 | 747 | 57 | |
D63485.1 | - | 6 - 7 | 866 | 57 |