Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRGPRX1
Entrez Gene ID
259249
Gene Name
MAS related GPR family member X1
Gene Aliases
GPCR, MGRG2, MRGX1, SNSR4
Chromosome Location
Chr.11: 18933813 - 18935002 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_147199.3 NP_671732.3 1 - 1 457 120
GenBank mRNA BC096835.1 - 1 - 1 465 120

View Full Product Details