Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location
Chr.1: 155946839 - 155991263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162383.1 NP_001155855.1 19 - 20 2375 86
NM_001162384.1 NP_001155856.1 19 - 20 2372 86
NM_004723.3 NP_004714.2 19 - 20 2300 86
XM_005245587.2 XP_005245644.1 19 - 20 2428 86
XM_005245588.2 XP_005245645.1 19 - 20 2425 86
XM_005245589.4 XP_005245646.1 19 - 20 2581 86
XM_005245590.3 XP_005245647.1 19 - 20 2303 86
XM_005245591.3 XP_005245648.1 20 - 21 2419 86
XM_005245592.2 XP_005245649.1 19 - 20 2572 86
XM_005245593.4 XP_005245650.1 19 - 20 2303 86
XM_005245594.2 XP_005245651.1 19 - 20 3289 86
XM_006711622.3 XP_006711685.1 19 - 20 3566 86
XM_006711623.3 XP_006711686.1 18 - 19 2302 86
XM_006711624.3 XP_006711687.1 18 - 19 2329 86
XM_006711625.3 XP_006711688.1 18 - 19 2235 86
XM_006711626.2 XP_006711689.1 18 - 19 3221 86
XM_011510136.2 XP_011508438.2 23 - 24 2984 86
XM_011510137.2 XP_011508439.2 23 - 24 2987 86
XM_011510139.2 XP_011508441.1 23 - 24 2987 86
XM_011510141.2 XP_011508443.1 19 - 20 3105 86
XM_011510143.2 XP_011508445.1 15 - 16 1851 86
XM_017002794.1 XP_016858283.1 23 - 24 2984 86
XM_017002795.1 XP_016858284.1 19 - 20 2319 86
XM_017002796.1 XP_016858285.1 19 - 20 3563 86
XM_017002797.1 XP_016858286.1 19 - 20 2303 86
XM_017002798.1 XP_016858287.1 18 - 19 3498 86
XM_017002799.1 XP_016858288.1 18 - 19 2299 86
GenBank mRNA AB014551.3 - 23 - 24 2643 86
AF486838.1 - 19 - 20 2254 86
AL512715.1 - 19 - 20 2285 86
BC020567.1 - 19 - 20 2357 86
BT007407.1 - 18 - 19 2176 86
U72206.1 - 19 - 20 2359 86
X15610.1 - 5 - 6 367 86

View Full Product Details