Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location
Chr.1: 155946839 - 155991263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162383.1 NP_001155855.1 6 - 7 693 67
XM_005245587.2 XP_005245644.1 6 - 7 746 67
XM_005245589.4 XP_005245646.1 6 - 7 899 67
XM_005245590.3 XP_005245647.1 6 - 7 621 67
XM_005245591.3 XP_005245648.1 7 - 8 737 67
XM_005245592.2 XP_005245649.1 6 - 7 890 67
XM_005245593.4 XP_005245650.1 6 - 7 621 67
XM_005245594.2 XP_005245651.1 6 - 7 1607 67
XM_006711622.3 XP_006711685.1 6 - 7 1884 67
XM_006711623.3 XP_006711686.1 5 - 6 620 67
XM_006711624.3 XP_006711687.1 5 - 6 647 67
XM_006711625.3 XP_006711688.1 5 - 6 553 67
XM_006711626.2 XP_006711689.1 5 - 6 1539 67
XM_011510137.2 XP_011508439.2 10 - 11 1305 67
XM_011510139.2 XP_011508441.1 10 - 11 1305 67
XM_011510141.2 XP_011508443.1 6 - 7 1423 67
XM_011510143.2 XP_011508445.1 2 - 3 169 67
XM_017002798.1 XP_016858287.1 5 - 6 1816 67
GenBank mRNA AK295924.1 - 6 - 7 890 67
AK304548.1 - 9 - 10 1030 67
BC020567.1 - 6 - 7 675 67
BT007407.1 - 5 - 6 494 67
U72206.1 - 6 - 7 677 67

View Full Product Details