The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152280.4 | NP_689493.3 | 1 - 2 | 334 | 72 |
XM_005245014.2 | XP_005245071.1 | 1 - 2 | 349 | 72 | |
XM_017000759.1 | XP_016856248.1 | 1 - 2 | 349 | 72 | |
GenBank mRNA | AK027540.1 | - | 1 - 2 | 236 | 72 |
AK074931.1 | - | 1 - 2 | 236 | 72 | |
BC004291.1 | - | 1 - 2 | 224 | 72 | |
BC013690.1 | - | 1 - 2 | 271 | 72 | |
BC039205.1 | - | 1 - 2 | 294 | 72 | |
BM801649.1 | - | 1 - 2 | 243 | 72 | |
CR749792.1 | - | 1 - 2 | 239 | 72 | |
D38522.2 | - | - | 263 | 72 | |
DA026743.1 | - | 1 - 2 | 263 | 72 | |
DA239014.1 | - | 1 - 2 | 334 | 72 |